Learn More
BURKLE INC Blue disposable powder funnel PS, Ø 3.94 in. (100 mm), sterile
Catalog No. 503081311
Encompass_Preferred
The blue disposable powder funnel, made of polystyrene, is produced in a cleanroom class 7 (10,000), individually packaged for single use, and sterilized.
The unique blue color doesn’t naturally occur in food, making it easy to spot during visual inspections if mixed into production.
Using blue tools supports HACCP hazard analysis by helping identify foreign objects, crucial in food, animal feed, and pharmaceutical industries.
The funnel features a short, wide discharge ideal for transferring powders, granulates, suspensions, and liquids.
Its wide opening prevents clogging, making it a practical, hygienic accessory for sampling and lab use.
When combined with filter paper, it is also suitable for filtering applications.
Catalog No. 50-308-1311 Supplier BURKLE INC Supplier No. 53786007
The blue disposable powder funnel, made of polystyrene, is produced in a cleanroom class 7 (10,000), individually packaged for single use, and sterilized.
The unique blue color doesn’t naturally occur in food, making it easy to spot during visual inspections if mixed into production.
Using blue tools supports HACCP hazard analysis by helping identify foreign objects, crucial in food, animal feed, and pharmaceutical industries.
The funnel features a short, wide discharge ideal for transferring powders, granulates, suspensions, and liquids.
Its wide opening prevents clogging, making it a practical, hygienic accessory for sampling and lab use.
When combined with filter paper, it is also suitable for filtering applications.