Learn More
Description
- 30 mL aqueous solution.
- Ideal for laboratory activity kits focusing on chromatography spectroscopy or dilution experiments.
- A consistent and reliable resource for hands-on science and research use.
Specifications
Specifications
| Chemical Name or Material | Blue #1 |
| CAS | 3844-45-9, 7732-18-5 |
| Color | Blue |
| pH | 5 to 8 |
| Quantity | 30 mL |
| Percent Purity | 0.5% |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.