Learn More
Alkali Scientific Blue Dye 20mL (Sterile) 20mL Vial, 10/Pk
SDP
Catalog No. 5000663174
Encompass_Preferred
Blue Dye, 20mL Sterile Vial, 10/Pk is a ready to use visualization solution designed to support aseptic process simulations, media fill trials, and operator qualification in controlled laboratory and cleanroom environments. The vivid blue coloration provides clear visual tracking of fluid movement, helping operators observe transfer accuracy and procedural performance during training and validation workflows. Packaged in sterile 20mL vials, the solution is suitable for use in pharmaceutical, biotechnology, and GMP regulated facilities where contamination control and process verification are essential. The dye enables realistic simulation of liquid handling without introducing microbial risk, making it a practical tool for validating aseptic technique, equipment setup, and fluid transfer procedures in quality assurance and cleanroom operations.
Catalog No. 50-006-63174 Supplier Alkali Scientific Supplier No. BDV000
Blue Dye, 20mL Sterile Vial, 10/Pk is a ready to use visualization solution designed to support aseptic process simulations, media fill trials, and operator qualification in controlled laboratory and cleanroom environments. The vivid blue coloration provides clear visual tracking of fluid movement, helping operators observe transfer accuracy and procedural performance during training and validation workflows. Packaged in sterile 20mL vials, the solution is suitable for use in pharmaceutical, biotechnology, and GMP regulated facilities where contamination control and process verification are essential. The dye enables realistic simulation of liquid handling without introducing microbial risk, making it a practical tool for validating aseptic technique, equipment setup, and fluid transfer procedures in quality assurance and cleanroom operations.