Learn More
Analytical Sales & Services Blue Fluorosilicone Mats for 48 Well (12mm OD) Reaction Blocks
Catalog No. 5000807040
Encompass
Blue Fluorosilicone Mats for 48 Well (12mm OD) Reaction Blocks\n\nQty/Pkg: Each
Catalog No. 50-008-07040 Supplier Analytical Sales & Services Supplier No. 48482F
Blue Fluorosilicone Mats for 48 Well (12mm OD) Reaction Blocks\n\nQty/Pkg: Each