Learn More
Research Products International Corp Bluo-Gal [5-Bromo-3-indolyl-B-D-galactopyranoside], 100 Milligrams
SDP
Supplier:  Research Products International Corp B725000.1
Encompass_Preferred
Chromogenic substrate for B-D-Galactosidase. Produces a deeper blue water insoluble color than X-GAL.
Catalog No. 50-213-194