Learn More
AMPLYUS LLC miniPCR bio blueGel™ electrophoresis with built-in transilluminator
SDP
Catalog No. NC1243149
Encompass
blueGel™ electrophoresis system with built-in transilluminator. Now 25% faster, brighter, and more portable! Results in 10 minutes. USB-C powered, compatible with portable batteries. Safe blue light illumination. Includes a silicone Fold-A-View™ photo documentation hood.
Catalog No. NC1243149 Supplier AMPLYUS LLC Supplier No. QP150001
blueGel™ electrophoresis system with built-in transilluminator. Now 25% faster, brighter, and more portable! Results in 10 minutes. USB-C powered, compatible with portable batteries. Safe blue light illumination. Includes a silicone Fold-A-View™ photo documentation hood.