Learn More
Enzo Life Sciences HIGHDEF blue IHC chromogen (HRP) (30 ml)
SDP
Supplier:  Enzo Life Sciences ADI9501510030
Encompass_Preferred
Unique high definition blue IHC chromogen for HRP-activated development. HIGHDEF blue IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridization (ISH) systems. Contents: HIGHDEF blue IHC chromogen (HRP) substrate buffer and chromogen, Empty mixing bottle. Application: IHC, ISH (in situ hybridization). Long Term Storage: +4°C
Catalog No. NC0868749