Learn More
Perkin Elmer US LLC Blue Injection Port Septa, Pkg. 10
Catalog No. 502112758
Encompass_Preferred
Septa rated to 275 °C. The blue septa are designed for routine applications. Employing a soft silicone rubber material and stay clean surface, they are easy to penetrate without a recess.
Catalog No. 50-211-2758 Supplier Perkin Elmer US LLC Supplier No. N9306873
Septa rated to 275 °C. The blue septa are designed for routine applications. Employing a soft silicone rubber material and stay clean surface, they are easy to penetrate without a recess.