Learn More
Cole-Parmer Keck Standard Taper Clips, Joint Size 19, Blue, Pk/12
Supplier:  Cole-Parmer 0684010
Encompass_Preferred
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
Catalog No. NC9907866