Learn More
Andwin Scientific BLUE LASER CRYO-BABIES LARG
SDP
Catalog No. 50821013
Encompass_Preferred
The Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading.
Catalog No. 50-821-013 Supplier Andwin Scientific Supplier No. 91871701
The Cryo-Babies labels can withstand most organic solvents, caustic agents, temperatures from -196C to 100C. This product can still be utilized while under oven, autoclaving, or extreme freezer temperatures. The labels can also be used on several surfaces like plastics, glass, and metals without cracking, peeling, or degrading.