Learn More
Andwin Scientific BLUE LIGHT AUTORAD FILM 8X10
SDP
Catalog No. NC9211251
Encompass_Preferred
The 8" x 10" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.
Catalog No. NC9211251 Supplier Andwin Scientific Supplier No. F9024
The 8" x 10" GeneMate blue autoradiography film is ideal for different applications like the chemiluminescence and autoradiography of 32P and 125I. This product lasts 2 years from the date of the manufacturing. It is sold in a box of 100.