Learn More
MTC Bio Blue Light Transilluminator with amber cover
Catalog No. NC1827680
Encompass_Preferred
Transilluminator, Blue Light, 465nm for use with safe green stains, 17x12cm viewing surface, 2 position removable amber cover, for viewing electrophoresis gels
Catalog No. NC1827680 Supplier MTC Bio Supplier No. TB4000
Transilluminator, Blue Light, 465nm for use with safe green stains, 17x12cm viewing surface, 2 position removable amber cover, for viewing electrophoresis gels