Learn More
MTC Bio SmartBlue UV viewing goggles, for blocking harmful UV rays from reaching eyes, 1/ea
Catalog No. NC1905723 Shop All Fisherbrand Products
Encompass_Preferred
SmartBlue Plus Viewing Glasses offer exceptional protection for researchers working with bright blue LED and UV light illuminators. The polycarbonate filter lenses are shatter proof, scratch resistant and block light in the UV (280nm) to blue (490nm) wavelengths. In addition, they are compliant with ANSI Z87.1.These glasses are ideal for general protection when working with hazardous wavelengths and for precision DNA band cutting on a UV or blue light transilluminator. While blocking hazardous UV and blue light, the glasses allow clear visualization of DNA bands stained with Ethidium Bromide, SmartGlow™, SYBR Green and other fluorescent dyes.
Catalog No. NC1905723
SmartBlue Plus Viewing Glasses offer exceptional protection for researchers working with bright blue LED and UV light illuminators. The polycarbonate filter lenses are shatter proof, scratch resistant and block light in the UV (280nm) to blue (490nm) wavelengths. In addition, they are compliant with ANSI Z87.1.These glasses are ideal for general protection when working with hazardous wavelengths and for precision DNA band cutting on a UV or blue light transilluminator. While blocking hazardous UV and blue light, the glasses allow clear visualization of DNA bands stained with Ethidium Bromide, SmartGlow™, SYBR Green and other fluorescent dyes.