Learn More
New England Biolabs, Inc. Blue Protein Loading Dye – 8 ml
SDP
Catalog No. 50811554
Encompass
The blue protein loading dye contains one vial of blue loading buffer and one vial of 30X reducing agent. The combined solution is ideal for protein gel applications.
Catalog No. 50-811-554 Supplier New England Biolabs, Inc. Supplier No. B7703S
Add to Cart
Add to Cart
The blue protein loading dye contains one vial of blue loading buffer and one vial of 30X reducing agent. The combined solution is ideal for protein gel applications.