Learn More
Biotium DNAzure Blue Nucleic Acid Gel Stain, 100X
Catalog No. NC1265435
Encompass_Preferred
DNAzure Blue Gel Stain allows you to visualize DNA bands in gels by eye, with detection sensitivity rivaling most fluorescence-based gel stains. Key to the technology is a DNA-binding dye that turns from colorless to deep blue upon exposure to light. The benefits include: deep blue bands visible by the naked eye following 5-30 min. light exposure, ultrasensitive detection (as little as ~1 ng DNA), and simplified DNA band excision without DNA-damaging UV Light.
Catalog No. NC1265435 Supplier Biotium Supplier No. 41020
DNAzure Blue Gel Stain allows you to visualize DNA bands in gels by eye, with detection sensitivity rivaling most fluorescence-based gel stains. Key to the technology is a DNA-binding dye that turns from colorless to deep blue upon exposure to light. The benefits include: deep blue bands visible by the naked eye following 5-30 min. light exposure, ultrasensitive detection (as little as ~1 ng DNA), and simplified DNA band excision without DNA-damaging UV Light.