Learn More
Bellco Glass, Inc. Blue Open Top S/Cap,32mm With 13/16 Hole"
SDP
Catalog No. 501945755
Encompass_Preferred
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Dimensions: 170x125
Catalog No. 50-194-5755 Supplier Bellco Glass, Inc. Supplier No. 563700035
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Dimensions: 170x125