Learn More
Chemglass Life Sciences Cap, GL-32, Blue, Open Top, (13/16" Dia Hole) Polypropylene
SDP
Catalog No. 501215137
Encompass
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5137 Supplier Chemglass Life Sciences Supplier No. CLS148120
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.