Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Blue Phantom Branched 4 Vessel Ultrasound Training Block Model
SDPSCI_ED

Catalog No. S00237 Shop All 3B Scientific Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S00237 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00237 Supplier 3B Scientific™ Supplier No. 3012457
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Branched 4 Vessel Ultrasound Training Block Model Includes four Vessels, Pre Filled with Red Ultrasound Fluid.

  • Branched 4 Vessel Ultrasound Training Block Model Includes:
    • Four Vessels (4 mm, 6 mm, and 8 mm Branched Vessels)
    • Pre Filled with Red Ultrasound Fluid

Specifications

Product Type Vessel Ultrasound Training Block
For Use With (Application) Ultrasound Training
Color Blue
Includes Four Vessels (4 mm, 6 mm, and 8 mm Branched Vessels)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.