Learn More
3B Scientific™ Blue Phantom TTE Ultrasound Training Platform Without Optional Head
SDPSCI_ED
Catalog No. S00544 Shop All 3B Scientific Products
Change view
Click to view available options
Training Platform; Includes: Microfiber Cleaning Towel, Powder, Syringes, and Ultrasound Fluid (Red, Blue, and Clear)
Catalog No.
S00544 Training Platform; Includes: Microfiber Cleaning Towel, Powder, Syringes, and Ultrasound Fluid (Red, Blue, and Clear)
1 options
This item is not returnable. View return policy
Catalog No. S00544 Supplier 3B Scientific™ Supplier No. 3012528
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Blue Phantom TTE Ultrasound Training Platform for Cardiac Procedures Including TTE and Pericardiocentesis.
  • Transthoracic Echocardiography (TTE) Ultrasound Training Platform Without optional Head Includes Full Body Torso (Mid Thigh to Upper Torso).
  • True Anatomy Series Ultrasound Training Platform for Cardiac Procedures Including TTE and Pericardiocentesis.
  • Ultrasound Anatomical Structures Include Heart Includes Left Ventricle, Right Ventricle, Left Atrium, and Right Atrium, Cardiac Valves, Left Atrial Appendage, Great Vessels, Pericardial Effusion.
  • Microfiber Cleaning Towel, Powder, Syringes, and Ultrasound Fluid (Red, Blue, and Clear)
Specifications
For Use With (Application) Cardiac Procedures Including TTE and Pericardiocentesis
Includes Microfiber Cleaning Towel, Powder, Syringes, and Ultrasound Fluid (Red, Blue, and Clear)
Product Type Training Platform