Learn More
Med Vet International BLue Plain 100% Plastic Micro Hematocrit Tubes, J0544P, 100/Tube, 10 Tubes/Box
Catalog No. 5000581777
Encompass_Preferred
Micro Hematocrit tubes. 100% Plastic. JorVet now offers 100% plastic microhematocrit capillary tubes that are virtually unbreakable. They eliminate the risk of injury and contamination associated with glass tubes due to breakage. Plastic microhematocrit.
Catalog No. 50-005-81777 Supplier Med Vet International Supplier No. JOR544P
Micro Hematocrit tubes. 100% Plastic. JorVet now offers 100% plastic microhematocrit capillary tubes that are virtually unbreakable. They eliminate the risk of injury and contamination associated with glass tubes due to breakage. Plastic microhematocrit.