Learn More
Cospheric LLC Blue Polyethylene Microspheres 1.00g/cc 250-300um - 10g
SDP
Catalog No. NC1714737
Encompass
Neutrally-buoyant blue polyethylene microspheres have a density of 1.00g/cc and can be used in flow visualization studies and particle image velocimetry (PIV). Highly opaque microspheres with maximum hiding power. Polymer microspheres with specific gravity of 1.00g/cc are designed to match the density of water for optimal suspension of particles, enabling visualization and characterization of fluid flow and testing the capability of devices to withstand particulate matter in the fluid stream.
Catalog No. NC1714737 Supplier Cospheric LLC Supplier No. BLPMS1.00 250300UM10G
Neutrally-buoyant blue polyethylene microspheres have a density of 1.00g/cc and can be used in flow visualization studies and particle image velocimetry (PIV). Highly opaque microspheres with maximum hiding power. Polymer microspheres with specific gravity of 1.00g/cc are designed to match the density of water for optimal suspension of particles, enabling visualization and characterization of fluid flow and testing the capability of devices to withstand particulate matter in the fluid stream.