Learn More
Bellco Glass, Inc. Blue Pouring Ring,32mm
SDP
Catalog No. 501945772
Encompass_Preferred
Provides drip free pouring and protects bottle rim from chipping. Autocavable Size: 32mm Color: blue
Catalog No. 50-194-5772 Supplier Bellco Glass, Inc. Supplier No. 563710032
Provides drip free pouring and protects bottle rim from chipping. Autocavable Size: 32mm Color: blue