Learn More
Cell Signaling Technology Blue Prestained Protein Marker, 11-250 kDa, 11 Bands, 250 μL
SDP
Catalog No. 502050821
Encompass
This product is a mixture of purified proteins covalently coupled to a blue dye that resolve into 11 bands ranging from 11 kDa to 250 kDa during electrophoresis. The protein concentrations are balanced for even intensity, and the dye coupling facilitates visualization during SDS-PAGE. The marker is supplied in a storage buffer containing 62.5 mM Tris-Phosphate (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol.
  • Provides 11 bands ranging from 11 kDa to 250 kDa.
  • Balanced protein concentration for even intensity.
  • Stable for 36 months when stored at -20°C.
  • Recommended for use with Tris-glycine SDS gels.
  • Important: do not boil the protein marker.
  • Requires thawing on ice and gentle vortexing before use.
Catalog No. 50-205-0821 Supplier Cell Signaling Technology Supplier No. 59329S
This product is a mixture of purified proteins covalently coupled to a blue dye that resolve into 11 bands ranging from 11 kDa to 250 kDa during electrophoresis. The protein concentrations are balanced for even intensity, and the dye coupling facilitates visualization during SDS-PAGE. The marker is supplied in a storage buffer containing 62.5 mM Tris-Phosphate (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol.
  • Provides 11 bands ranging from 11 kDa to 250 kDa.
  • Balanced protein concentration for even intensity.
  • Stable for 36 months when stored at -20°C.
  • Recommended for use with Tris-glycine SDS gels.
  • Important: do not boil the protein marker.
  • Requires thawing on ice and gentle vortexing before use.