Learn More
Perkin Elmer US LLC Blue, PSS Splitless deactivated glass liners
Catalog No. 502104650
Encompass_Preferred
PSS splitless deactivated glass liners 1.25 mm I.D. x 4.0 O.D. x 86.2, with deactivated wool 5/pack.
Catalog No. 50-210-4650 Supplier Perkin Elmer US LLC Supplier No. N9306237
PSS splitless deactivated glass liners 1.25 mm I.D. x 4.0 O.D. x 86.2, with deactivated wool 5/pack.