Learn More
Heathrow Scientific™ Blue and Purple Reagent Reservoir Chiller
Description
- Reagents are kept at less than 5°C (41°F) for approximately 3 hours
- Ice-free solution eliminates the risk of contamination from messy ice baths
- Stackable, save space when storing in the freezer or on the bench top
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
- Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
- Made from sonic welded, engineered polypropylene that ensures a leak free design
- Not intended for autoclave, do not use with liquid nitrogen
- Reusable
- Two-year warranty
Specifications
Specifications
| Capacity (English) | 1.7 oz. |
| Color | Blue and Purple |
| Capacity (Metric) | 50 mL |
| Dimensions (L x W x H) | 5.70 x 3 x 1.80 in. |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Type | Reservoir Cooler |
| Quantity | 2 Pk. |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.