Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Blue and Purple Reagent Reservoir Chiller

Catalog No. 03449277
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-449-277 100 mL
03-449-274 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-449-277 Supplier Heathrow Scientific™ Supplier No. 120735
Add to Cart
Add to Cart

Reagent Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.

  • Reagents are kept at less than 5°C (41°F) for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Stackable, save space when storing in the freezer or on the bench top
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Made from sonic welded, engineered polypropylene that ensures a leak free design
  • Not intended for autoclave, do not use with liquid nitrogen
  • Reusable
  • Two-year warranty

Specifications

Capacity (English) 3.4 oz.
Color Blue and Purple
Capacity (Metric) 100 mL
Dimensions (L x W x H) 5.90 x 3.50 x 1.80 in.
Material Polypropylene
Sterility Non-sterile
Type Reservoir Cooler
Quantity 2 Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Reusable
Bottom Shape Convoluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.