Learn More
Bellco Glass, Inc. Blue S/Cap W/PTFE/RR Septa,9mm 9-425mm
SDP
Catalog No. 501945384
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 9mm
Catalog No. 50-194-5384 Supplier Bellco Glass, Inc. Supplier No. 207000900
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 9mm