Learn More
Bioworld BLUE SEPAROPOREAGAROSE) 4
Supplier:  Bioworld 201810963
Encompass
Blue Separopore (Agarose) 4B-CL. Technical Specifications: Ligand: Cibacron Blue F3G-A; Matrix: Separopore 4B-CL (crosslinked agarose beads, 4%); Particle size range: 52 - 165 um; Matrix activation: Triazine coupling; Ligand density: 2 - 6 umol Cibacron blue F3G-A / ml drained gel; Binding capacity: ~15 mg bovine serum albumin / ml drained gel; pH stability: 3 - 13; Storage: 2 - 8 deg.C; Supplied as 50% slurry in 0.5M NaCl containing 0.02% thimerosal. 100ml.
Catalog No. 50-197-910