Learn More
Bioworld BLUE SEPAROPOREAGAROSE) 4
Supplier: Bioworld 201810961
This item has a minimum order quantity of 3 per supplier requirements. BLUE SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix for purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins.The ligand, Cibacron Blue F3G-A, is covalently coupled t...