Learn More
Bioworld BLUE SEPAROPOREAGAROSE) 4
Catalog No. 50197909
Encompass
BLUE SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix for purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins.The ligand, Cibacron Blue F3G-A, is covalently coupled t...
Catalog No. 50-197-909 Supplier Bioworld Supplier No. 201810962
BLUE SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix for purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins.The ligand, Cibacron Blue F3G-A, is covalently coupled t...