Learn More
Bioworld BLUE SEPAROPOREAGAROSE) 6
Supplier:  Bioworld 201810262
Encompass
BLUE SEPAROPORE (AGAROSE) 6B-CL: Blue Separopore CL-6B A Cibacron blue dye-ligand affinity matrix for purification of albumin, enzymes (including NAD+ and NADP+), coagulation factors, interferons and related proteins. The ligand, Ciba...
Catalog No. 50-197-728