Learn More
Bioworld BLUE SEPAROPOREAGAROSE) 6
Supplier:  Bioworld 201810261
Encompass
Blue Separopore (Agarose) 6B-CL. Technical Specifications: Ligand: Cibacron Blue F3G-A; Matrix: Separopore 6B-CL (crosslinked agarose, 6%); Particle size range: 52 - 165 um; Matrix activation: Triazine coupling; Coupling conditions: pH 9 - 10; Ligand density: 8 umol; Cibacron Blue/ml drained gel; Binding capacity: > 15 - 20 mg bovine serum albumin/ml drained gel; pH stability: 3 - 13; Chemical stability: Stable to all commonly used aqueous buffers and additives such as 8M urea and 6M guanidine hydrochloride.Physical stability: Negligible volume variation due to changes in pH or ionic strength.Supplied as Suspension in 0.5M NaCl containing 0.02% thimerosal; Storage: 2 - 8 deg.C. 25ml.
Catalog No. 50-197-727