Learn More
Sigma Aldrich Fine Chemicals Biosciences Blue SepharoseR 6 fast flo
Catalog No. 502904270
Encompass
Blue Sepharose(R) 6 fast flow is Cibacron Blue 3G coupled to Sepharose(R) 6 fast flow. This medium is well established as adsorbent for albumin and interferon at both laboratory and process scale.Blue Sepharose(R) 6 fast flow is a member of the Cytiva range of affinity chromatography media for capture and intermediate purification. Blue Sepharose(R) 6 fast flow is composed of cross-linked 6% agarose beads modified with Cibacron Blue 3G covalently attached by the triazine coupling method. The blue dye binds many proteins such as albumin interferon lipoproteins and blood coagulation factors. It also binds several enzymes including kinases dehydrogenases and most enzymes requiring adenyl-containing cofactors e.g. NAD.Blue Sepharose(R) 6 fast flow has been developed and tested in co-operation with leading large scale manufacturers and is used in routine commercial production.Blue Sepharose(R) 6 fast flow is available in a range of different bulk pack sizes and convenient pre-packed fo
Catalog No. 50-290-4270 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17094801
Blue Sepharose(R) 6 fast flow is Cibacron Blue 3G coupled to Sepharose(R) 6 fast flow. This medium is well established as adsorbent for albumin and interferon at both laboratory and process scale.Blue Sepharose(R) 6 fast flow is a member of the Cytiva range of affinity chromatography media for capture and intermediate purification. Blue Sepharose(R) 6 fast flow is composed of cross-linked 6% agarose beads modified with Cibacron Blue 3G covalently attached by the triazine coupling method. The blue dye binds many proteins such as albumin interferon lipoproteins and blood coagulation factors. It also binds several enzymes including kinases dehydrogenases and most enzymes requiring adenyl-containing cofactors e.g. NAD.Blue Sepharose(R) 6 fast flow has been developed and tested in co-operation with leading large scale manufacturers and is used in routine commercial production.Blue Sepharose(R) 6 fast flow is available in a range of different bulk pack sizes and convenient pre-packed fo