Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Blue SMS Coverall with Attached Hood, Elastic Wrists and Ankles

Catalog No. 17184199 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-184-199 2X-Large
17-184-194 Medium
17-184-195 Large
17-184-193 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-184-199 Supplier International Enviroguard™ Supplier No. 2225B2XL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMS coveralls provide three layers of protection against particulates such as dirt and dust, and light liquid splashes or sprays. Lightweight and breathable fabric allows heat to escape so wearers stay cool.

  • SMS coveralls are lightweight and breathable, so wearers stay cool throughout long shifts
  • Durable, three-layer SMS fabric (Spunbond Meltblown Spunbond) protects against light liquid splashes and sprays and provides an ideal particulate barrier
  • Storm flap covers the zipper to help keep non-hazardous particles and liquids from entering zipper seams
  • Wider, gusseted leg area gives wearer more room to move in all directions without ripping fabric
  • Attached hood along with elastic wrists and ankles offer a secure fit and prevent skin exposure
  • Elastic back stretches with user for more comfort and better tear resistance
  • Possible applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color Royal Blue
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle
Latex Free Yes
Model 2225B
No. of Pockets None
For Use With (Application) Asbestos and Lead Abatement, Mold Remediation, Manufacturing, Assembly Operations
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.