Learn More
Leica Biosystems Leica Blue Snowcoat Microscope Slides, 1 x 3 in x 0.04 in, pack of 1,440
Catalog No. NC3237836
Encompass
Leica Blue Snowcoat Microscope Slides are standard 1 x 3 in (25.4 x 76.2 mm), 0.04 in thick slides with a blue Snowcoat marking area for color-coded sample identification and durable labeling. Sold in packs of 1,440, they accept xylene-resistant marking pens and pencils for routine histology and laboratory identification.
  • Standard 1 x 3 in microscope slide, 0.04 in thickness.
  • Blue Snowcoat marking area for easy color-coded identification.
  • Compatible with xylene-resistant marking pens or pencils.
  • Pack contains 1,440 slides for high-volume use.
  • Intended for routine histology and laboratory labeling.
Catalog No. NC3237836 Supplier Leica Biosystems Supplier No. 3800320
Leica Blue Snowcoat Microscope Slides are standard 1 x 3 in (25.4 x 76.2 mm), 0.04 in thick slides with a blue Snowcoat marking area for color-coded sample identification and durable labeling. Sold in packs of 1,440, they accept xylene-resistant marking pens and pencils for routine histology and laboratory identification.
  • Standard 1 x 3 in microscope slide, 0.04 in thickness.
  • Blue Snowcoat marking area for easy color-coded identification.
  • Compatible with xylene-resistant marking pens or pencils.
  • Pack contains 1,440 slides for high-volume use.
  • Intended for routine histology and laboratory labeling.