Learn More
Braintree Scientific Blue Spandex Rodent Jacket, Rat - XLarge, 500+ grams
SDP
Catalog No. 501955702
Encompass
Made of double layers of Lycra/Spandex fabric with Velcro fasteners for adjustment and secure fit. Available in 5 standard sizes, (X-small, Small, Medium, Large and X-large), order by weight range of your animals. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5702 Supplier Braintree Scientific Supplier No. RJHXL
Made of double layers of Lycra/Spandex fabric with Velcro fasteners for adjustment and secure fit. Available in 5 standard sizes, (X-small, Small, Medium, Large and X-large), order by weight range of your animals. **Sold for research purposes only and not for human use**