Learn More
Thermco Products Inc BLUE SPIRIT THERM OS62C
Catalog No. NC1290241
Encompass_Preferred
ASTM S62C thermometer (Blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.
Catalog No. NC1290241 Supplier Thermco Products Inc Supplier No. ACC062CBLSFC
ASTM S62C thermometer (Blue spirit) certified at the ASTM recommended temperatures and calibrated against N.I.S.T. standards. A N.I.S.T. traceable document is completed in the Thermco ISO17025/ILAC accedited laboratory.