Learn More
Leica Biosystems Leica Biosystems Blue Super Cassettes, 100/Case - 3 x 2 x ¾ in (75 x 52 x 17 mm)
Catalog No. NC3711073
Encompass
Super cassettes are large-format histology embedding and processing cassettes designed to accommodate oversized specimens such as large brain sections, eyes, intact joints, whole organs, whole prostates, and multiple-tissue cases. They are intended for use in veterinary, research, and pharmaceutical laboratories; processing schedules may need to be adjusted for larger specimens.
  • Holds large or intact specimens for embedding and processing.
  • Case contains 100 loose cassettes for bulk use.
  • Color: blue, for easy specimen identification.
  • Dimensions: 3 x 2 x ¾ in (75 x 52 x 17 mm).
  • Suitable for veterinary, research, and pharmaceutical histology workflows.
  • Processing schedules may need to be adjusted for larger specimens.
Catalog No. NC3711073 Supplier Leica Biosystems Supplier No. 38VSP59062
Super cassettes are large-format histology embedding and processing cassettes designed to accommodate oversized specimens such as large brain sections, eyes, intact joints, whole organs, whole prostates, and multiple-tissue cases. They are intended for use in veterinary, research, and pharmaceutical laboratories; processing schedules may need to be adjusted for larger specimens.
  • Holds large or intact specimens for embedding and processing.
  • Case contains 100 loose cassettes for bulk use.
  • Color: blue, for easy specimen identification.
  • Dimensions: 3 x 2 x ¾ in (75 x 52 x 17 mm).
  • Suitable for veterinary, research, and pharmaceutical histology workflows.
  • Processing schedules may need to be adjusted for larger specimens.