Learn More
Med Vet International Blue Suture Wing, 4.5fr, J1112A, 10/Pack
Catalog No. 5000583582
Encompass_Preferred
7fr blue suture wing. NG tube or line catheter through channel bax material. Non-sterile, 2 suture holes. Pebax material.
Catalog No. 50-005-83582 Supplier Med Vet International Supplier No. JOR1112A
7fr blue suture wing. NG tube or line catheter through channel bax material. Non-sterile, 2 suture holes. Pebax material.