Learn More
Medchemexpress LLC Blue Tetrazolium | 1871-22-3 | 99.8% | 1 G
SDP
Catalog No. 5000411036
Encompass_Preferred
Blue Tetrazolium is a blue dye used in microbial research. It can be reduced into blue tetrazolium formazan (BTF) and is suitable for determining the activity of succinate dehydrogenase (SDH) in yeast strains. The BTF can be extracted from cells using DMSO, and its absorption spectrum can be tested, showing a wide wavelength range of 480-600 nm with maximal absorbance at 540 nm.
  • Blue dye for microbial research.
  • Can be reduced into blue tetrazolium formazan (BTF).
  • Used to determine the activity of succinate dehydrogenase (SDH) in yeast strains.
  • BTF exhibits a wide wavelength range of 480-600 nm with maximal absorbance at 540 nm.
  • Combines with succinate dehydrogenase (SDH).
Catalog No. 50-004-11036 Supplier Medchemexpress LLC Supplier No. HYD00071G
Blue Tetrazolium is a blue dye used in microbial research. It can be reduced into blue tetrazolium formazan (BTF) and is suitable for determining the activity of succinate dehydrogenase (SDH) in yeast strains. The BTF can be extracted from cells using DMSO, and its absorption spectrum can be tested, showing a wide wavelength range of 480-600 nm with maximal absorbance at 540 nm.
  • Blue dye for microbial research.
  • Can be reduced into blue tetrazolium formazan (BTF).
  • Used to determine the activity of succinate dehydrogenase (SDH) in yeast strains.
  • BTF exhibits a wide wavelength range of 480-600 nm with maximal absorbance at 540 nm.
  • Combines with succinate dehydrogenase (SDH).