Learn More
Medchemexpress LLC Blue Tetrazolium | 1871-22-3 | MFCD00040933 | 99.8% | 100 MG
SDP
Catalog No. 5000411038
Encompass_Preferred
Blue Tetrazolium is a blue dye used in microbial research that can be reduced into blue tetrazolium formazan (BTF). It acts as a substrate to determine the activity of succinate dehydrogenase (SDH) in yeast strains. BTF can be extracted from cells using DMSO, and its absorption spectrum can be analyzed with a broad wavelength range of 480-600 nm, and a maximal absorbance at 540 nm.
  • Used as a blue dye in microbial research.
  • Reducible to blue tetrazolium formazan (BTF).
  • Determines succinate dehydrogenase (SDH) activity in yeast strains.
  • Absorption spectrum of BTF shows maximal absorbance at 540 nm.
  • Functions as a novel corrosion inhibitor for cold-rolled steel in hydrochloric acid solutions.
Catalog No. 50-004-11038 Supplier Medchemexpress LLC Supplier No. HYD0007100MG
Blue Tetrazolium is a blue dye used in microbial research that can be reduced into blue tetrazolium formazan (BTF). It acts as a substrate to determine the activity of succinate dehydrogenase (SDH) in yeast strains. BTF can be extracted from cells using DMSO, and its absorption spectrum can be analyzed with a broad wavelength range of 480-600 nm, and a maximal absorbance at 540 nm.
  • Used as a blue dye in microbial research.
  • Reducible to blue tetrazolium formazan (BTF).
  • Determines succinate dehydrogenase (SDH) activity in yeast strains.
  • Absorption spectrum of BTF shows maximal absorbance at 540 nm.
  • Functions as a novel corrosion inhibitor for cold-rolled steel in hydrochloric acid solutions.