Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Blue tetrazolium (tetrazolium blue) | 1871-22-3 | 99.8% | 500 MG
SDP

Catalog No. 5000410805
Encompass_Preferred

Blue Tetrazolium (Tetrazolium blue) is a blue dye used for microbial research. It can be reduced into blue tetrazolium formazan (BTF). Blue Tetrazolium can be used to determine the activity of succinate dehydrogenase (SDH) in yeast strains, where it is reduced as a substrate. DMSO can be used to extract BTF from cells, and the absorption spectrum of BTF can be tested. BTF exhibits a wide wavelength range of 480-600 nm with a maximal absorbance at 540 nm. Blue Tetrazolium combines with succinate dehydrogenase (SDH).

  • Molecular weight: 727.64
  • Formula: C40H32Cl2N8O2
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: RT, sealed storage, away from moisture and light. In solvent: -80°C for 1 year, -20°C for 6 months

Catalog No. 50-004-10805 Supplier Medchemexpress LLC Supplier No. HYD0007500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blue Tetrazolium (Tetrazolium blue) is a blue dye used for microbial research. It can be reduced into blue tetrazolium formazan (BTF). Blue Tetrazolium can be used to determine the activity of succinate dehydrogenase (SDH) in yeast strains, where it is reduced as a substrate. DMSO can be used to extract BTF from cells, and the absorption spectrum of BTF can be tested. BTF exhibits a wide wavelength range of 480-600 nm with a maximal absorbance at 540 nm. Blue Tetrazolium combines with succinate dehydrogenase (SDH).

  • Molecular weight: 727.64
  • Formula: C40H32Cl2N8O2
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: RT, sealed storage, away from moisture and light. In solvent: -80°C for 1 year, -20°C for 6 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.