Learn More
Caresfield™ Blue Textured Tourniquet
SDP
Catalog No. 21222048
Change view
Click to view available options
Packaging Type:
Bulk Rolled
Dispenser Box
Flat Pack
Individually Rolled
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
21-222-048 Flat Pack
21-222-047 Dispenser Box
21-222-040 Pack
21-222-049 Individually Rolled
21-222-050 Bulk Rolled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 21-222-048 Supplier Caresfield™ Supplier No. 48TXBFP
Add to Cart
Add to Cart
This item is not returnable. View return policy
Disposable, non-latex tourniquets designed with patient safety and comfort in mind.
  • Disposable
  • Smooth material
  • Bright colors
  • Odor-free and powder-free
  • Easy dispensing
Specifications
Color Blue
Latex Free Yes
Quantity 250/Pk., 4/Cs.
Product Type Adult Tourniquet
Disposable Single-use
Dimensions (L x W) 1 x 18 in.
Packaging Type Flat Pack