Learn More
Andwin Scientific BLUE TOUGH-SPOTS SMALL 3/8IN
SDP
Supplier:  Andwin Scientific 91850001
Encompass_Preferred
The blue Tough-Spots labels are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels.
Catalog No. 50-820-979