Learn More
Quality Biological Inc Permanent Blue Ink Tube Marker, Fast Drying Blue Ink, 5/CS
SDP
Catalog No. 50147441
Encompass_Preferred
Permanent Blue Ink Tube Marker, Fast Drying Blue Ink, 5/CS
Fast drying, smudge resistant, fine point marker
Ideal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes!
Alcohol/Solvent Resistant: No
Catalog No. 50-147-441 Supplier Quality Biological Inc Supplier No. P886229406
Permanent Blue Ink Tube Marker, Fast Drying Blue Ink, 5/CS
Fast drying, smudge resistant, fine point marker
Ideal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes!
Alcohol/Solvent Resistant: No