Learn More
MSC MSA V-Gard Slotted Cap, Type 1 Class E, 1 ea
Catalog No. 502886363
Encompass
This V-Gard Slotted Cap is an impact-resistant hard hat designed for head protection. It is classified as Type 1 and Class E, offering reliable safety. The cap features a 4-point suspension system and is constructed from durable polyethylene.
  • V-Gard Slotted Cap style for enhanced protection
  • Standard type with impact-resistant application
  • Blue, non-vented design with cap brim style
  • Constructed from durable polyethylene material
  • ANSI Type I and Class E rated for safety standards
  • Features a 4-point plastic suspension with ratchet adjustment
  • Accommodates head sizes from 6-1/2 to 8
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards
Catalog No. 50-288-6363 Supplier MSC Supplier No. 98722663
This V-Gard Slotted Cap is an impact-resistant hard hat designed for head protection. It is classified as Type 1 and Class E, offering reliable safety. The cap features a 4-point suspension system and is constructed from durable polyethylene.
  • V-Gard Slotted Cap style for enhanced protection
  • Standard type with impact-resistant application
  • Blue, non-vented design with cap brim style
  • Constructed from durable polyethylene material
  • ANSI Type I and Class E rated for safety standards
  • Features a 4-point plastic suspension with ratchet adjustment
  • Accommodates head sizes from 6-1/2 to 8
  • Complies with ANSI/ISEA Z89.1-2014 and CSA Z94.1-2005 standards