Learn More
Tewes Corporation 18 x 24 4 Mil Vci Flat Bag
SDP
Catalog No. NC1841700
Encompass_Preferred
VCI Bags (Vapor Corrosion Inhibitors) are easy to use and provide cost effective proven corrosion protection. Multi-Metal Blue bags are a special packaging film containing proprietary Zerust corrosion inhibiting additives developed to prevent the corrosion of parts that contain combinations of ferrous and non-ferrous metals in typical shipping and storage conditions. Use Zerust Multi-Metal Film to protect electronic equipment & components, machinery, and any other part composed of multiple metals. Bags will protect metals for up to 5 years if properly packed and stored. Offer a shelf life of 2 years if stored at 85 degrees F (30 degrees C) or less, out of direct sunlight and in original packaging. 250/CASE
Catalog No. NC1841700 Supplier Tewes Corporation Supplier No. 025M00169
VCI Bags (Vapor Corrosion Inhibitors) are easy to use and provide cost effective proven corrosion protection. Multi-Metal Blue bags are a special packaging film containing proprietary Zerust corrosion inhibiting additives developed to prevent the corrosion of parts that contain combinations of ferrous and non-ferrous metals in typical shipping and storage conditions. Use Zerust Multi-Metal Film to protect electronic equipment & components, machinery, and any other part composed of multiple metals. Bags will protect metals for up to 5 years if properly packed and stored. Offer a shelf life of 2 years if stored at 85 degrees F (30 degrees C) or less, out of direct sunlight and in original packaging. 250/CASE