Learn More
Electron Microscopy Sciences Blue Wax Dissection Tray, 5/PK
SDP
Catalog No. 502090705
Encompass_Preferred
A dissection tray with a wax base pinning surface for detailed dissections. These trays are specially formulated to allow pins to stick firmly and be easily removed. They are made using a completely environmentally safe wax product, instead of the "black wax" product used in most dissection trays. Blue wax contains no toxic chemicals, and no fire, health or reactivity hazards are associated with it. Blue wax can be heated to renew the surface and the dissection tray can be reused several times.
Catalog No. 50-209-0705 Supplier Electron Microscopy Sciences Supplier No. 6236605
A dissection tray with a wax base pinning surface for detailed dissections. These trays are specially formulated to allow pins to stick firmly and be easily removed. They are made using a completely environmentally safe wax product, instead of the "black wax" product used in most dissection trays. Blue wax contains no toxic chemicals, and no fire, health or reactivity hazards are associated with it. Blue wax can be heated to renew the surface and the dissection tray can be reused several times.