Learn More
Sigma Aldrich Fine Chemicals Biosciences Blue White SelectTM Screen
Catalog No. 502905191
Encompass
Sigmas Blue-White Select(TM) Screening Reagent is a chromogenic substrate for beta-galactosidase used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Blue-White Select(TM) Screening Reagent is designed for blue-white selection of recombinant bacterial colonies with the lac phenotype.
Catalog No. 50-290-5191 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B39285ML
Sigmas Blue-White Select(TM) Screening Reagent is a chromogenic substrate for beta-galactosidase used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Blue-White Select(TM) Screening Reagent is designed for blue-white selection of recombinant bacterial colonies with the lac phenotype.