Learn More
AMPLYUS LLC miniPCR bio blueBox™ Pro Transilluminator with Imaging Hood
SDP
Catalog No. NC1897743
Encompass_Preferred
blueBox™ Pro transilluminator with foldable darkroom, allowing you to take high quality gel images with your phone. Adjustable hinged lid for easy band excision; compatible with GelGreen, SeeGreen™, and SYBR family DNA stains.
Catalog No. NC1897743 Supplier AMPLYUS LLC Supplier No. QP170003
blueBox™ Pro transilluminator with foldable darkroom, allowing you to take high quality gel images with your phone. Adjustable hinged lid for easy band excision; compatible with GelGreen, SeeGreen™, and SYBR family DNA stains.